Learn More
Abcam Tigecycline, Glycylcycline antibiotic, 25MG
Catalog No. 502706378
Encompass_Preferred
MW 585.6 Da, Purity >99%. Glycylcycline antibiotic. Active against Gram-negative and Gram-positive bacteria. Active in vivo. Active against multi-drug resistant pathogens.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6378 Supplier Abcam Supplier No. AB14200025MG
MW 585.6 Da, Purity >99%. Glycylcycline antibiotic. Active against Gram-negative and Gram-positive bacteria. Active in vivo. Active against multi-drug resistant pathogens.
The product is subject to the following: Abcam Restricted Use Statement