Learn More
Abcam Tigecycline, Glycylcycline antibiotic, 5MG
Catalog No. 502706377
Encompass_Preferred
MW 585.6 Da, Purity >99%. Glycylcycline antibiotic. Active against Gram-negative and Gram-positive bacteria. Active in vivo. Active against multi-drug resistant pathogens.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6377 Supplier Abcam Supplier No. AB1420005MG
MW 585.6 Da, Purity >99%. Glycylcycline antibiotic. Active against Gram-negative and Gram-positive bacteria. Active in vivo. Active against multi-drug resistant pathogens.
The product is subject to the following: Abcam Restricted Use Statement