Learn More
Medchemexpress LLC HY-B0117C 10mg Medchemexpress, Tigecycline (tetramesylate) CAS: Purity:>98%
SDP
Catalog No. 501932902
Encompass_Preferred
Medchemexpress, HY-B0117C 10mg Tigecycline (tetramesylate) CAS: Tigecycline tetramesylate (GAR-936 tetramesylate) is a broad-spectrum glycylcycline antibiotic. The mean inhibitory concentration (MIC) of Tigecycline for E. coli (MG1655 strain) is approximately 125 ng/mL [1] . MIC 50 and MIC 90 are 1 and 2 mg/L for Acinetobacter baumannii (A. baumannii), respectively [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2902 Supplier Medchemexpress LLC Supplier No. HYB0117C10MG
Medchemexpress, HY-B0117C 10mg Tigecycline (tetramesylate) CAS: Tigecycline tetramesylate (GAR-936 tetramesylate) is a broad-spectrum glycylcycline antibiotic. The mean inhibitory concentration (MIC) of Tigecycline for E. coli (MG1655 strain) is approximately 125 ng/mL [1] . MIC 50 and MIC 90 are 1 and 2 mg/L for Acinetobacter baumannii (A. baumannii), respectively [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.