Learn More
Medchemexpress LLC Tigecycline tetramesylate | 97.0% | 5 MG
SDP
Catalog No. 5000450914
Encompass_Preferred
Tigecycline tetramesylate is a broad-spectrum glycylcycline antibiotic effective against various bacterial strains, including *E. coli* and *Acinetobacter baumannii*. It has shown inhibitory effects on AML2 and HL-60 cells in vitro and has demonstrated the ability to reduce tumor volume in acute myeloid leukemia xenograft models in vivo. This compound is suitable for diverse research applications requiring antibiotic properties.
  • Broad-spectrum glycylcycline antibiotic
  • Soluble in DMSO, H2O, and PBS
  • Requires ultrasonication for optimal dissolution in certain solvents
  • Solid storage at 4°C, sealed, away from moisture
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C
Catalog No. 50-004-50914 Supplier Medchemexpress LLC Supplier No. HYB0117C5MG
Tigecycline tetramesylate is a broad-spectrum glycylcycline antibiotic effective against various bacterial strains, including *E. coli* and *Acinetobacter baumannii*. It has shown inhibitory effects on AML2 and HL-60 cells in vitro and has demonstrated the ability to reduce tumor volume in acute myeloid leukemia xenograft models in vivo. This compound is suitable for diverse research applications requiring antibiotic properties.
  • Broad-spectrum glycylcycline antibiotic
  • Soluble in DMSO, H2O, and PBS
  • Requires ultrasonication for optimal dissolution in certain solvents
  • Solid storage at 4°C, sealed, away from moisture
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C