Learn More
Medchemexpress LLC Tigecycline tetramesylate | 97.0% | 50 MG
SDP
Catalog No. 5000450861
Encompass_Preferred
Tigecycline tetramesylate is a broad-spectrum glycylcycline antibiotic. It demonstrates a mean inhibitory concentration (MIC) of approximately 125 ng/mL for E. coli (MG1655 strain), with MIC50 and MIC90 values of 1 mg/L and 2 mg/L, respectively, for Acinetobacter baumannii.
  • Broad-spectrum glycylcycline antibiotic
  • Mean inhibitory concentration (MIC) of 125 ng/mL for E. coli
  • Inhibits AML2 and HL-60 cells
  • Reduces tumor volume and weight in NOD/SCID mice
Catalog No. 50-004-50861 Supplier Medchemexpress LLC Supplier No. HYB0117C50MG
Tigecycline tetramesylate is a broad-spectrum glycylcycline antibiotic. It demonstrates a mean inhibitory concentration (MIC) of approximately 125 ng/mL for E. coli (MG1655 strain), with MIC50 and MIC90 values of 1 mg/L and 2 mg/L, respectively, for Acinetobacter baumannii.
  • Broad-spectrum glycylcycline antibiotic
  • Mean inhibitory concentration (MIC) of 125 ng/mL for E. coli
  • Inhibits AML2 and HL-60 cells
  • Reduces tumor volume and weight in NOD/SCID mice