Learn More
Medchemexpress LLC Tigecycline (tetramesylate) | 97.01% | 100 MG
SDP
Catalog No. 5000450873
Encompass_Preferred
Tigecycline tetramesylate is a broad-spectrum glycylcycline antibiotic with a purity of 97.01%. It has a molecular formula of C33H55N5O20S4 and a molecular weight of 970.07. This light yellow to yellow solid is effective against bacteria such as *E. coli* and *Acinetobacter baumannii*.
  • Broad-spectrum glycylcycline antibiotic
  • Effective against *E. coli* and *A. baumannii*
  • Suitable for in vitro and in vivo studies
  • Light yellow to yellow solid appearance
  • Stored at 4°C, sealed, away from moisture
  • In solvent, stored at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-50873 Supplier Medchemexpress LLC Supplier No. HYB0117C100MG
Tigecycline tetramesylate is a broad-spectrum glycylcycline antibiotic with a purity of 97.01%. It has a molecular formula of C33H55N5O20S4 and a molecular weight of 970.07. This light yellow to yellow solid is effective against bacteria such as *E. coli* and *Acinetobacter baumannii*.
  • Broad-spectrum glycylcycline antibiotic
  • Effective against *E. coli* and *A. baumannii*
  • Suitable for in vitro and in vivo studies
  • Light yellow to yellow solid appearance
  • Stored at 4°C, sealed, away from moisture
  • In solvent, stored at -80°C for 6 months or -20°C for 1 month