Learn More
MSC North Tigerhood Classic Welding Helmet, Thermoplastic, Shade 10, 1 ea
Catalog No. 5000507738
Encompass
The Tigerhood™ Classic lift front welding helmet is designed for professional welding applications, providing reliable protection and comfort. Constructed from durable thermoplastic, this gray helmet features a unique airflow design that helps block fumes and smoke from the wearer's face while offering extended throat protection. The helmet includes a shade 10 filter plate and is equipped with a free-floating headgear system that allows for multiple adjustments to ensure an individualized fit.
  • Exclusive airflow design blocks fumes and smoke while extending throat protection
  • Molded-in color prevents chipping, peeling, and fading
  • Includes shade 10 filter plate, clear cover plate, and mounting system
  • Free-floating headgear with multiple adjustment points
  • Supports integration with hard hats via mounting mechanism
  • Lens size: 2 in. x 4 1/4 in.
Catalog No. 50-005-07738 Supplier MSC Supplier No. 93281335
The Tigerhood™ Classic lift front welding helmet is designed for professional welding applications, providing reliable protection and comfort. Constructed from durable thermoplastic, this gray helmet features a unique airflow design that helps block fumes and smoke from the wearer's face while offering extended throat protection. The helmet includes a shade 10 filter plate and is equipped with a free-floating headgear system that allows for multiple adjustments to ensure an individualized fit.
  • Exclusive airflow design blocks fumes and smoke while extending throat protection
  • Molded-in color prevents chipping, peeling, and fading
  • Includes shade 10 filter plate, clear cover plate, and mounting system
  • Free-floating headgear with multiple adjustment points
  • Supports integration with hard hats via mounting mechanism
  • Lens size: 2 in. x 4 1/4 in.