Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGIT Detection Set, ProSci
SDP

Catalog No. 16595252 Shop All ProSci Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-252 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-252 Supplier ProSci LLC Supplier No. SD8800SET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Llama Monoclonal Detection Sets

TIGIT Monoclonal specifically detects TIGIT in Human samples. It is validated for ELISA, IHC-P, ICC, IF, Flow

TIGIT Antibody: The T cell immunoreceptor with Ig and ITIM domains (TIGIT) is a member of the PVR (poliovirus receptor) family of immunoglobin proteins. It is expressed on several classes of T cells including follicular B helper T cells (TFH). TIGIT has been shown to bind PVR with high affinity; this binding is thought to assist interactions between TFH and dendritic cells to regulate T cell dependent B cell responses (1). Similar to other immune checkpoint proteins such as PD-1, TIGIT is upregulated on exhausted T cells in chronic viral infections and cancer. Blockade of both TIGIT and PD-1 pathways leads to tumor rejection in mice suggesting that it may be of therapeutic use against cancer (2).

Specifications

Antigen TIGIT
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Flow Cytometry
Classification Monoclonal
Concentration Antibody 1 mg/mL
Conjugate Unconjugated
Formulation TIGIT Antibodies are supplied in PBS.
Host Species Llama
Purification Method TIGIT Antibodies are affinity chromatography purified via Nickel column. Antibodies are supplied as a His-tagged purified protein. They also contains a myc-tag for detection.
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage TIGIT antibodies should be stored in working aliquots at -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Detection Sets
Form Liquid
Isotype Not Applicable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.