Learn More
ACROBiosystems ACROBiosystems Rat TIGIT Protein, Fc Tag
Supplier:  ACROBiosystems TITR52581MG
Encompass_Preferred
Recombinant Protein;1MG;Rat TIGIT, Fc Tag (TIT-R5258) is expressed from human 293 cells (HEK293). It contains AA Ala 17 - Ala 138 (Accession # D3ZTQ2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 39.9 kDa. As a result of glycosylation, the protein migrates as 48-55 kDa under reducing (R) condition, and 85-110 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human Nectin-2, His Tag (Cat. No. PV2-H52E2) at 5 μg/mL (100 μL/well) can bind Rat TIGIT, Fc Tag (Cat. No. TIT-R5258) with a linear range of 0.039-5 μg/mL (QC tested).
Catalog No. 50-195-7410