Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque TIGIT Protein, His Tag (HPLC-verified)
Catalog No. 501957123
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque TIGIT, His Tag (HPLC-verified) (TIT-C5223) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Accession # XP_015300911.1). In the region Met 89 - Ile 208, the AA sequence of Cynomolgus and Rhesus macaque TIGIT are homologus.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.2 kDa. The protein migrates as 18-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque TIGIT, His Tag (HPLC-verified) (Cat. No. TIT-C5223) at 5 μg/mL (100 μL/well) can bind Human CD155, Fc Tag (Cat. No. CD5-H5251) with a linear range of 0.078-2.5 μg/mL (QC tested).
Catalog No. 50-195-7123 Supplier ACROBiosystems Supplier No. TITC52231MG
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque TIGIT, His Tag (HPLC-verified) (TIT-C5223) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Accession # XP_015300911.1). In the region Met 89 - Ile 208, the AA sequence of Cynomolgus and Rhesus macaque TIGIT are homologus.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.2 kDa. The protein migrates as 18-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque TIGIT, His Tag (HPLC-verified) (Cat. No. TIT-C5223) at 5 μg/mL (100 μL/well) can bind Human CD155, Fc Tag (Cat. No. CD5-H5251) with a linear range of 0.078-2.5 μg/mL (QC tested).