Learn More
ACROBiosystems ACROBiosystems Mouse TIGIT Protein, His Tag
Catalog No. 501957288
Encompass_Preferred
Recombinant Protein;1MG;Mouse TIGIT, His Tag (TIT-M52E6) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession # NP_001139797.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.8 kDa. The protein migrates as 18-30 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Mouse TIGIT, His Tag (Cat. No. TIT-M52E6) at 5 μg/mL (100 μL/well) can bind Mouse CD155, Mouse IgG2a Fc Tag (Cat. No. CD5-M5254) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-195-7288 Supplier ACROBiosystems Supplier No. TITM52E61MG
Recombinant Protein;1MG;Mouse TIGIT, His Tag (TIT-M52E6) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession # NP_001139797.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.8 kDa. The protein migrates as 18-30 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Mouse TIGIT, His Tag (Cat. No. TIT-M52E6) at 5 μg/mL (100 μL/well) can bind Mouse CD155, Mouse IgG2a Fc Tag (Cat. No. CD5-M5254) with a linear range of 5-78 ng/mL (QC tested).