Learn More
ACROBiosystems ACROBiosystems Canine TIGIT Protein, His Tag
Catalog No. 501956783
Encompass_Preferred
Recombinant Protein;100UG;Canine TIGIT, His Tag (TIT-C52H7) is expressed from human 293 cells (HEK293). It contains AA Phe 17 - Pro 208 (Accession # F1PKJ0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.1 kDa. The protein migrates as 25-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Canine TIGIT, His Tag (Cat. No. TIT-C52H7) at 5 μg/mL (100 μL/well) can bind Human CD155, mouse IgG2a Fc tag, low endotoxin (Cat. No. CD5-H5254) with a linear range of 0.01-0.313 μg/mL (QC tested).
Catalog No. 50-195-6783 Supplier ACROBiosystems Supplier No. TITC52H7100UG
Recombinant Protein;100UG;Canine TIGIT, His Tag (TIT-C52H7) is expressed from human 293 cells (HEK293). It contains AA Phe 17 - Pro 208 (Accession # F1PKJ0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.1 kDa. The protein migrates as 25-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Canine TIGIT, His Tag (Cat. No. TIT-C52H7) at 5 μg/mL (100 μL/well) can bind Human CD155, mouse IgG2a Fc tag, low endotoxin (Cat. No. CD5-H5254) with a linear range of 0.01-0.313 μg/mL (QC tested).