Learn More
Sino Biological Recombinant Human TIGIT Protein (hFc Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10917H02HB100
Encompass_Preferred
A DNA sequence encoding the TIGIT (NP_776160.2) (Met1-Pro141) was expressed with the Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6095