Learn More
ACROBiosystems Recombinant Protein;Mouse TIGIT Protein, Fc Tag;HEK293;1MG;TIT,TIGIT,VSIG9,VSTM3,VSIG-9,VSTM-3
Catalog No. 501658678
Encompass_Preferred
Recombinant Protein;Mouse TIGIT Protein, Fc Tag;1MG;Mouse TIGIT, Fc Tag (TIT-M5257) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession # NP_001139797).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40 kDa. The protein migrates as 45-52 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Mouse CD155, mouse IgG2a Fc tag (Cat. No. CD5-M5254) at 2 μg/mL (100 μL/well) can bind Mouse TIGIT, Fc Tag (Cat. No. TIT-M5257) with a linear range of 1-16 ng/mL (QC tested).
Catalog No. 50-165-8678 Supplier ACROBiosystems Supplier No. TITM52571MG
Recombinant Protein;Mouse TIGIT Protein, Fc Tag;1MG;Mouse TIGIT, Fc Tag (TIT-M5257) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession # NP_001139797).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40 kDa. The protein migrates as 45-52 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Mouse CD155, mouse IgG2a Fc tag (Cat. No. CD5-M5254) at 2 μg/mL (100 μL/well) can bind Mouse TIGIT, Fc Tag (Cat. No. TIT-M5257) with a linear range of 1-16 ng/mL (QC tested).