Learn More
Selleck Chemical LLC Tiglic acid S3789-25mg
SDP
Catalog No. 503108993
Encompass_Preferred
Tiglic acid (Cevadic acid Sabadillic acid Tiglinic acid) is a monocarboxylic unsaturated organic acid found in croton oil and in several other natural products It is used in making perfumes and flavoring agents
Catalog No. 50-310-8993 Supplier Selleck Chemical LLC Supplier No. S378925MG
Tiglic acid (Cevadic acid Sabadillic acid Tiglinic acid) is a monocarboxylic unsaturated organic acid found in croton oil and in several other natural products It is used in making perfumes and flavoring agents