Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tiglic acid | 80-59-1 | 100.0% | 100.12 | 25 G
SDP

Catalog No. 5000451397
Encompass_Preferred

Tiglic acid is a monocarboxylic unsaturated organic acid that can be isolated from croton oil. It agonizes FFA2, upregulates PYY expression, and its derivatives have anti-inflammatory activity. It is used in the research of obesity, diabetes, and inflammatory diseases.

  • Appearance: Solid, white to light yellow
  • Structure classification: Ketones, aldehydes, acids
  • Initial source: Plants Verbenaceae Lantana camara Linn.
  • Endogenous metabolite: Yes
  • Targets: Free Fatty Acid Receptor, Neuropeptide Y Receptor
  • Pathway: GPCR/G Protein, neuronal signaling
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 2 years, -20°C for 1 year
  • Solubility (in vitro): DMSO 100 mg/mL, H2O 5 mg/mL

Catalog No. 50-004-51397 Supplier Medchemexpress LLC Supplier No. 25GHYW012999
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tiglic acid is a monocarboxylic unsaturated organic acid that can be isolated from croton oil. It agonizes FFA2, upregulates PYY expression, and its derivatives have anti-inflammatory activity. It is used in the research of obesity, diabetes, and inflammatory diseases.

  • Appearance: Solid, white to light yellow
  • Structure classification: Ketones, aldehydes, acids
  • Initial source: Plants Verbenaceae Lantana camara Linn.
  • Endogenous metabolite: Yes
  • Targets: Free Fatty Acid Receptor, Neuropeptide Y Receptor
  • Pathway: GPCR/G Protein, neuronal signaling
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 2 years, -20°C for 1 year
  • Solubility (in vitro): DMSO 100 mg/mL, H2O 5 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.