Learn More
Medchemexpress LLC Tiglic acid | 80-59-1 | 99.97% | 100.12 | 5 G
SDP
Catalog No. 5000452172
Encompass_Preferred
Tiglic acid is a monocarboxylic unsaturated organic acid that can be isolated from croton oil. It agonizes FFA2 and upregulates PYY expression. Tiglic acid derivatives have anti-inflammatory activity, making it suitable for research into obesity, diabetes, and inflammatory diseases.
  • Agonizes FFA2
  • Upregulates PYY expression
  • Derivatives show anti-inflammatory activity
  • Useful in obesity research
  • Useful in diabetes research
  • Useful in inflammatory diseases research
Catalog No. 50-004-52172 Supplier Medchemexpress LLC Supplier No. HYW0129995G
Tiglic acid is a monocarboxylic unsaturated organic acid that can be isolated from croton oil. It agonizes FFA2 and upregulates PYY expression. Tiglic acid derivatives have anti-inflammatory activity, making it suitable for research into obesity, diabetes, and inflammatory diseases.
  • Agonizes FFA2
  • Upregulates PYY expression
  • Derivatives show anti-inflammatory activity
  • Useful in obesity research
  • Useful in diabetes research
  • Useful in inflammatory diseases research