Learn More
Cayman Chemical TIgloylLcarnItInchLRIde 10mg
SDP
Catalog No. 502989774
Encompass_Preferred
A short-chain acylcarnitine; plasma levels are increased in several melanoma mouse xenograft models compared with non-tumor bearing mice; levels are increased in dried blood spots from neonates with T2 deficiency, an inborn error of isoleucine metabolism characterized by ketoacidosis
Catalog No. 50-298-9774 Supplier Cayman Chemical Supplier No. 3099410 MG
A short-chain acylcarnitine; plasma levels are increased in several melanoma mouse xenograft models compared with non-tumor bearing mice; levels are increased in dried blood spots from neonates with T2 deficiency, an inborn error of isoleucine metabolism characterized by ketoacidosis