Learn More
Medchemexpress LLC Tiglyl carnitine | 64681-36-3 | MFCD31657519 | 99.9% | 243.30 g/mol | C12H21NO4 | 5 MG
SDP
Catalog No. 501969250
Encompass_Preferred
Tiglyl carnitine (CAS 64681-36-3) is provided as an analytical reference standard for research and analytical workflows. It is intended for use in metabolite identification, method development, and mass spectrometry quantitation. A certificate of analysis and analytical data are supplied for each batch to support traceability and method validation.
  • High purity analytical standard suitable for LC-MS and GC-MS analyses.
  • Supplied in small, weighed quantities for analytical workflows.
  • Includes certificate of analysis with batch-specific assay and HPLC results.
  • Soluble in DMSO; prepare solutions according to laboratory best practices.
  • Intended for metabolite identification, biomarker validation, and method development.
  • Store and handle under conditions recommended in the SDS.
Catalog No. 50-196-9250 Supplier Medchemexpress LLC Supplier No. HY1134085MG
Tiglyl carnitine (CAS 64681-36-3) is provided as an analytical reference standard for research and analytical workflows. It is intended for use in metabolite identification, method development, and mass spectrometry quantitation. A certificate of analysis and analytical data are supplied for each batch to support traceability and method validation.
  • High purity analytical standard suitable for LC-MS and GC-MS analyses.
  • Supplied in small, weighed quantities for analytical workflows.
  • Includes certificate of analysis with batch-specific assay and HPLC results.
  • Soluble in DMSO; prepare solutions according to laboratory best practices.
  • Intended for metabolite identification, biomarker validation, and method development.
  • Store and handle under conditions recommended in the SDS.