Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tiglyl carnitine | 64681-36-3 | 99.8% | 243.30 | 1 ML
SDP

Catalog No. 5000436861
Encompass_Preferred

Tiglyl carnitine is an endogenous metabolite linked to celiac disease and mitochondrial acetoacetyl-CoA thiolase (T2) deficiency. It presents as a white to off-white solid and is categorized under Ketones, Aldehydes, and Acids. It is intended for research use only.

  • Soluble in DMSO at 25 mg/mL (102.75 mM).
  • Detailed protocols for preparing stock solutions and in vivo dissolution are provided.
  • Storage recommendations: -20°C for 3 years (powder) and -80°C for 6 months or -20°C for 1 month (in solvent).

Catalog No. 50-004-36861 Supplier Medchemexpress LLC Supplier No. HY11340810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tiglyl carnitine is an endogenous metabolite linked to celiac disease and mitochondrial acetoacetyl-CoA thiolase (T2) deficiency. It presents as a white to off-white solid and is categorized under Ketones, Aldehydes, and Acids. It is intended for research use only.

  • Soluble in DMSO at 25 mg/mL (102.75 mM).
  • Detailed protocols for preparing stock solutions and in vivo dissolution are provided.
  • Storage recommendations: -20°C for 3 years (powder) and -80°C for 6 months or -20°C for 1 month (in solvent).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.