Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tiglyl carnitine | 64681-36-3 | MFCD31657519 | 99.9% | 243.30 g/mol | C12H21NO4 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY113408R5MG

Encompass_Preferred

Tiglyl carnitine is an analytical standard used as a reference material for metabolic and biochemical research. Supplied as a solid, it is characterized by molecular formula C12H21NO4 and molecular weight 243.30 g/mol, and is offered in small quantities suitable for analytical applications.

  • Purity 99.9% for analytical accuracy.
  • Molecular weight 243.30 g/mol.
  • Molecular formula C12H21NO4.
  • Supplied as a solid in small analytical quantities (e.g., 5 mg).
  • Intended for use as an analytical standard in metabolic and biochemical studies.
  • Soluble in DMSO for solution preparation.

Catalog No. 50-002-64184


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.