Learn More
Medchemexpress LLC Tiglyl carnitine (standard) | 64681-36-3 | 98.0% | 243.30 | 10 MG
SDP
Catalog No. 5000436574
Encompass_Preferred
Tiglyl carnitine is an analytical standard intended for research and analytical applications. It is associated with celiac disease and mitochondrial acetoacetyl-CoA thiolase (T2) deficiency.
  • Analytical standard
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-36574 Supplier Medchemexpress LLC Supplier No. HY113408R10MG
Tiglyl carnitine is an analytical standard intended for research and analytical applications. It is associated with celiac disease and mitochondrial acetoacetyl-CoA thiolase (T2) deficiency.
  • Analytical standard
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS