Learn More
Medchemexpress LLC Tigulixostat | 1287766-55-5 | 99.2% | C16H14N4O2 | 10MG
SDP
Catalog No. 5000259156
Encompass_Preferred
Tigulixostat is an orally active, non-purine selective xanthine oxidase inhibitor used in preclinical and biochemical research to investigate uric acid metabolism and potential treatments for gout and hyperuricemia. It is provided as a solid suitable for preparation of stock solutions for in vitro and in vivo studies.
  • High purity (99.2%).
  • Non-purine selective xanthine oxidase inhibitor.
  • Suitable for biochemical and preclinical research applications.
  • Provided as a solid for solution preparation.
  • Available in small research pack sizes for convenience.
Catalog No. 50-002-59156 Supplier Medchemexpress LLC Supplier No. HY13958510MG
Tigulixostat is an orally active, non-purine selective xanthine oxidase inhibitor used in preclinical and biochemical research to investigate uric acid metabolism and potential treatments for gout and hyperuricemia. It is provided as a solid suitable for preparation of stock solutions for in vitro and in vivo studies.
  • High purity (99.2%).
  • Non-purine selective xanthine oxidase inhibitor.
  • Suitable for biochemical and preclinical research applications.
  • Provided as a solid for solution preparation.
  • Available in small research pack sizes for convenience.