Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tigulixostat | 1287766-55-5 | MFCD11977724 | 95.0% | 294.31 g·mol^-1 | C16H14N4O2 | 5 MG
SDP

Catalog No. 5000221148
Encompass_Preferred

Tigulixostat is an orally active, non-purine selective xanthine oxidase inhibitor used in preclinical research to reduce serum uric acid in studies of gout and hyperuricemia. It is supplied as a powder and is suitable for solution preparation for biochemical and cellular assays.

  • Selective xanthine oxidase inhibition with low-nanomolar potency.
  • Supplied as a powder for laboratory use.
  • Molecular formula C16H14N4O2; molecular weight 294.31 g·mol⁻¹.
  • Pack size provided as 5 MG with solution concentration preparations noted.
  • Storage: powder -20°C; in solvent -80°C up to 6 months, -20°C up to 1 month.

Catalog No. 50-002-21148 Supplier Medchemexpress LLC Supplier No. HY1395855MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tigulixostat is an orally active, non-purine selective xanthine oxidase inhibitor used in preclinical research to reduce serum uric acid in studies of gout and hyperuricemia. It is supplied as a powder and is suitable for solution preparation for biochemical and cellular assays.

  • Selective xanthine oxidase inhibition with low-nanomolar potency.
  • Supplied as a powder for laboratory use.
  • Molecular formula C16H14N4O2; molecular weight 294.31 g·mol⁻¹.
  • Pack size provided as 5 MG with solution concentration preparations noted.
  • Storage: powder -20°C; in solvent -80°C up to 6 months, -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.