Learn More
Medchemexpress LLC Tildacerfont | 1014983-00-6 | 98.6% | 419.97 | 250 MG
SDP
Catalog No. 5000439506
Encompass_Preferred
Tildacerfont is a potent and orally active corticotrophin-releasing factor type 1 (CRF1) receptor antagonist. It effectively reduces adrenocorticotropic hormone (ACTH) and adrenal androgen levels. Tildacerfont has a favorable safety profile and can be used for researching congenital adrenal hyperplasia.
  • Potent and orally active CRF1 receptor antagonist.
  • Effectively reduces ACTH and adrenal androgen levels.
  • Favorable safety profile.
  • Can be used for researching congenital adrenal hyperplasia.
Catalog No. 50-004-39506 Supplier Medchemexpress LLC Supplier No. 250MGHY113603
Tildacerfont is a potent and orally active corticotrophin-releasing factor type 1 (CRF1) receptor antagonist. It effectively reduces adrenocorticotropic hormone (ACTH) and adrenal androgen levels. Tildacerfont has a favorable safety profile and can be used for researching congenital adrenal hyperplasia.
  • Potent and orally active CRF1 receptor antagonist.
  • Effectively reduces ACTH and adrenal androgen levels.
  • Favorable safety profile.
  • Can be used for researching congenital adrenal hyperplasia.