Learn More
Cayman Chemical TIldIpIrosIn 10mg
SDP
Catalog No. 502979973
Encompass_Preferred
A 16-membered macrolide used as an antibiotic in veterinary medicine; inhibits protein synthesis in bacteria and blocks the production of biofilms; particularly effective against Gram-negative pathogens (MIC = 0.25-1 UG/ml against P. multocida and M. haemolytica)
Catalog No. 50-297-9973 Supplier Cayman Chemical Supplier No. 1708910 MG
A 16-membered macrolide used as an antibiotic in veterinary medicine; inhibits protein synthesis in bacteria and blocks the production of biofilms; particularly effective against Gram-negative pathogens (MIC = 0.25-1 UG/ml against P. multocida and M. haemolytica)