Learn More
Selleck Chemical LLC Tildipirosin
SDP
Catalog No. 5000002992
Encompass_Preferred
Tildipirosin is a 16-membered ring tribasic and semi-synthetic macrolide antibiotic
Catalog No. 50-000-02992 Supplier Selleck Chemical LLC Supplier No. S64232MG
Tildipirosin is a 16-membered ring tribasic and semi-synthetic macrolide antibiotic