Learn More
Medchemexpress LLC HY-N2555 5mg Medchemexpress, Tilianin CAS:4291-60-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN25555MG
Encompass_Preferred
Medchemexpress, HY-N2555 5mg Tilianin CAS:4291-60-5 Tilianin is an active flavonoid glycoside found in many medical plants, with potential anti-hypertensive, myocardial-protective, anti-diabetic, anti-hyperlipidemic, anti-inflammatory and antioxidant effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2903