Learn More
TARGETMOL CHEMICALS INC TILIQUINOL 25MG
SDP
Catalog No. 502579373
Encompass_Preferred
Also available in 50 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Tiliquinol (NSC-130828) (NSC-130828) is a non-absorbed luminal agent with anti-amoebic properties that is often used in conjunction with tibroquinol. It is commonly employed in the study and investigation of amebic liver abscesses. purity: 99%
Catalog No. 50-257-9373 Supplier TARGETMOL CHEMICALS INC Supplier No. T6435225MG
Also available in 50 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Tiliquinol (NSC-130828) (NSC-130828) is a non-absorbed luminal agent with anti-amoebic properties that is often used in conjunction with tibroquinol. It is commonly employed in the study and investigation of amebic liver abscesses. purity: 99%