Learn More
Medchemexpress LLC Tiliquinol | 5541-67-3 | MFCD00864652 | 99.6% | 159.18 g/mol | C10H9NO | 250 MG
SDP
Catalog No. 5000302490
Encompass_Preferred
Tiliquinol is a non-absorbed anti-amoebic luminal agent (NSC 130828) used in research applications, including studies of amebic liver abscess and investigations of combination therapies. It is provided in solid form and as a DMSO solution, with associated purity data and safety documentation for laboratory use.
  • Non-absorbed anti-amoebic luminal agent for in vitro and in vivo research.
  • Used in combination therapies to study amebic liver abscess.
  • Available as solid powder and as a DMSO solution for dosing flexibility.
  • High purity suitable for research applications.
  • Safety data sheet and technical documentation available for laboratory handling.
Catalog No. 50-003-02490 Supplier Medchemexpress LLC Supplier No. 250MGHYW018397
Tiliquinol is a non-absorbed anti-amoebic luminal agent (NSC 130828) used in research applications, including studies of amebic liver abscess and investigations of combination therapies. It is provided in solid form and as a DMSO solution, with associated purity data and safety documentation for laboratory use.
  • Non-absorbed anti-amoebic luminal agent for in vitro and in vivo research.
  • Used in combination therapies to study amebic liver abscess.
  • Available as solid powder and as a DMSO solution for dosing flexibility.
  • High purity suitable for research applications.
  • Safety data sheet and technical documentation available for laboratory handling.