Learn More
MSC Tillman Anti-Vibration MIG Welding Gloves, Premium Cowhide, Medium, 1 ea
Catalog No. 5000687880
Encompass
These anti-vibration MIG welding gloves are engineered to reduce hand fatigue and prevent vibration-related injuries for professional welders. They are meticulously crafted with a premium top grain cowhide palm and a rugged split cowhide back, ensuring professional-grade durability and protection.
  • Anti-vibration gel technology
  • Premium cowhide construction
  • Cotton fleece lined for comfort and insulation
  • Elastic back design for snug fit and natural hand movement
  • Kevlar® reinforced stitching for seam strength
  • Extended 4 in. cuff for forearm protection
  • Vibration-reducing palm pads
  • Professional MIG approved
  • Ergonomic comfort design
Catalog No. 50-006-87880 Supplier MSC Supplier No. 58425968
These anti-vibration MIG welding gloves are engineered to reduce hand fatigue and prevent vibration-related injuries for professional welders. They are meticulously crafted with a premium top grain cowhide palm and a rugged split cowhide back, ensuring professional-grade durability and protection.
  • Anti-vibration gel technology
  • Premium cowhide construction
  • Cotton fleece lined for comfort and insulation
  • Elastic back design for snug fit and natural hand movement
  • Kevlar® reinforced stitching for seam strength
  • Extended 4 in. cuff for forearm protection
  • Vibration-reducing palm pads
  • Professional MIG approved
  • Ergonomic comfort design