Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R3 Safety™ Tillman™ Premium Elkskin Stick Welding Gloves

Catalog No. 19819962 Shop All R3 Safety Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-819-962 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-819-962 Supplier R3 Safety™ Supplier No. 750L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed of premium elkskin leather that stays soft even when it gets hot. The back is lined with cotton/foam for additional heat protection, while the palm is unlined for enhanced dexterity.

  • Pearl premium elkskin stays soft even when hot
  • Durable stitching with DuPont™ Aramid Fibers for additional strength
  • Welted fingers protect stitching
  • Cotton/foam lined back for added heat protection
  • Unlined palm for enhanced dexterity
  • Extended, 14 in. gauntlet cuff
  • Reinforced thumb

Specifications

Color Gray
Grade Premium
Hand Specific
Material Elkskin Leather
Size Large
Style Gunn
Glove Cuff Style Gauntlet
Liner Material Cotton, Foam
Series 750
Thumb Style Straight
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.