Learn More
MSC TILLMAN Flame Resistant/Retardant Welding Jacket, Large, Cotton, Snaps, 1 ea
Catalog No. 5000685386
Encompass
This flame resistant and retardant welding jacket is designed for protective use in welding applications. Made from cotton, it offers comfort and safety with a snap closure.
  • Designed for MIG welding and TIG welding applications
  • Provides protection against accidental flame
  • Meets ASTM F1506 standards
  • Features a 9 oz material weight
Catalog No. 50-006-85386 Supplier MSC Supplier No. 71279830
This flame resistant and retardant welding jacket is designed for protective use in welding applications. Made from cotton, it offers comfort and safety with a snap closure.
  • Designed for MIG welding and TIG welding applications
  • Provides protection against accidental flame
  • Meets ASTM F1506 standards
  • Features a 9 oz material weight