Learn More
MSC Tillman Welding Jacket, Flame Resistant, Cotton, 2X-Large, Green, 1 each
Catalog No. 5000685388
Encompass
This flame resistant cotton jacket offers protection against sudden flame exposure, light welding, and sparks. Made from 9 oz. flame-retardant cotton, it features a 30 in. length and snap front closure.
  • Flame resistant 100% cotton construction
  • Protects against sudden flame and sparks
  • Provides 30 in. length for extended coverage
  • Inside pocket for storage
  • Washable fabric meets ASTM D6413-10 standard
  • Snap fasteners for secure closure
  • Suitable for light duty welding applications
  • Maintains flame resistance for up to 50 home or 25 commercial washings
Catalog No. 50-006-85388 Supplier MSC Supplier No. 71279897
This flame resistant cotton jacket offers protection against sudden flame exposure, light welding, and sparks. Made from 9 oz. flame-retardant cotton, it features a 30 in. length and snap front closure.
  • Flame resistant 100% cotton construction
  • Protects against sudden flame and sparks
  • Provides 30 in. length for extended coverage
  • Inside pocket for storage
  • Washable fabric meets ASTM D6413-10 standard
  • Snap fasteners for secure closure
  • Suitable for light duty welding applications
  • Maintains flame resistance for up to 50 home or 25 commercial washings