Learn More
Medchemexpress LLC HY-U00197 1mg Medchemexpress, Tilmacoxib CAS:180200-68-4 Purity:>98%
SDP
Catalog No. 502083191
Encompass_Preferred
Medchemexpress, HY-U00197 1mg Tilmacoxib CAS:180200-68-4 Tilmacoxib (JTE522) is a highly selective, time-dependent and irreversible human COX-2 inhibitor with an IC50 of 85 nM in an enzyme assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3191 Supplier Medchemexpress LLC Supplier No. HYU001971MG
Medchemexpress, HY-U00197 1mg Tilmacoxib CAS:180200-68-4 Tilmacoxib (JTE522) is a highly selective, time-dependent and irreversible human COX-2 inhibitor with an IC50 of 85 nM in an enzyme assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.