Learn More
Medchemexpress LLC Tilmicosin | 108050-54-0 | 98.0% | 100 MG
SDP
Catalog No. 502082670
Encompass_Preferred
Tilmicosin is an orally active calcium channel antagonist and macrolide antibiotic with antimicrobial activity. It primarily acts on the 50S subunit of bacterial ribosomes, inhibiting protein synthesis. Tilmicosin is effective in treating respiratory diseases in livestock such as cattle, sheep, and pigs, and also has immunomodulatory and anti-inflammatory effects.
  • Acts on the 50S subunit of bacterial ribosomes, inhibiting protein synthesis.
  • Effective in treating respiratory diseases in livestock.
  • Exhibits immunomodulatory effects.
  • Exhibits anti-inflammatory effects.
  • Has an MIC for 90% of Staphylococcus aureus isolates from bovine udder of 0.78 μg/mL.
  • Induces apoptosis of bovine neutrophils.
  • Inhibits the expression of COX-2 and iNOS in LPS-treated macrophages.
Catalog No. 50-208-2670 Supplier Medchemexpress LLC Supplier No. HYB0905100MG
Tilmicosin is an orally active calcium channel antagonist and macrolide antibiotic with antimicrobial activity. It primarily acts on the 50S subunit of bacterial ribosomes, inhibiting protein synthesis. Tilmicosin is effective in treating respiratory diseases in livestock such as cattle, sheep, and pigs, and also has immunomodulatory and anti-inflammatory effects.
  • Acts on the 50S subunit of bacterial ribosomes, inhibiting protein synthesis.
  • Effective in treating respiratory diseases in livestock.
  • Exhibits immunomodulatory effects.
  • Exhibits anti-inflammatory effects.
  • Has an MIC for 90% of Staphylococcus aureus isolates from bovine udder of 0.78 μg/mL.
  • Induces apoptosis of bovine neutrophils.
  • Inhibits the expression of COX-2 and iNOS in LPS-treated macrophages.