Learn More
Cayman Chemical Tilmicosin-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2714486-61-8, 1 mg
SDP
Catalog No. 502949802
Encompass_Preferred
An internal standard for the quantification of tilmicosin by GC- or LC-MS
Catalog No. 50-294-9802 Supplier Cayman Chemical Supplier No. 335451 MG
An internal standard for the quantification of tilmicosin by GC- or LC-MS