Learn More
Medchemexpress LLC Tilmicosin-d3 | 2714486-61-8 | 5 MG
SDP
Catalog No. 5000408734
Encompass_Preferred
Tilmicosin-d3 is the deuterium labeled Tilmicosin. Tilmicosin (LY-177370) is an orally active calcium channel antagonist and macrolide antibiotic with antimicrobial activity. It primarily acts on the 50S subunit of bacterial ribosomes, inhibiting protein synthesis.
  • Effective in treating respiratory diseases in livestock such as cattle, sheep, and pigs.
  • Possesses immunomodulatory and anti-inflammatory effects.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08734 Supplier Medchemexpress LLC Supplier No. HYB0905S5MG
Tilmicosin-d3 is the deuterium labeled Tilmicosin. Tilmicosin (LY-177370) is an orally active calcium channel antagonist and macrolide antibiotic with antimicrobial activity. It primarily acts on the 50S subunit of bacterial ribosomes, inhibiting protein synthesis.
  • Effective in treating respiratory diseases in livestock such as cattle, sheep, and pigs.
  • Possesses immunomodulatory and anti-inflammatory effects.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.