Learn More
Medchemexpress LLC Tilmicosin (phosphate) | 137330-13-3 | 98.0% | 500 MG
SDP
Catalog No. 5000408984
Encompass_Preferred
Tilmicosin phosphate (LY-177370 phosphate) is an orally active macrolide antibiotic and calcium channel antagonist. This compound exhibits significant antimicrobial activity by inhibiting protein synthesis through its action on the 50S subunit of bacterial ribosomes. It is widely used in veterinary medicine for the treatment of respiratory diseases in livestock, such as cattle, sheep, and pigs, and has also been noted for its immunomodulatory and anti-inflammatory effects.
  • Orally active macrolide antibiotic
  • Functions as a calcium channel antagonist
  • Inhibits bacterial protein synthesis via the 50S ribosomal subunit
  • Effective against respiratory diseases in livestock
  • Possesses immunomodulatory properties
  • Demonstrates anti-inflammatory effects
  • Recommended storage at 4°C, sealed and away from moisture. In solution, store at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-004-08984 Supplier Medchemexpress LLC Supplier No. HYB0905A500MG
Tilmicosin phosphate (LY-177370 phosphate) is an orally active macrolide antibiotic and calcium channel antagonist. This compound exhibits significant antimicrobial activity by inhibiting protein synthesis through its action on the 50S subunit of bacterial ribosomes. It is widely used in veterinary medicine for the treatment of respiratory diseases in livestock, such as cattle, sheep, and pigs, and has also been noted for its immunomodulatory and anti-inflammatory effects.
  • Orally active macrolide antibiotic
  • Functions as a calcium channel antagonist
  • Inhibits bacterial protein synthesis via the 50S ribosomal subunit
  • Effective against respiratory diseases in livestock
  • Possesses immunomodulatory properties
  • Demonstrates anti-inflammatory effects
  • Recommended storage at 4°C, sealed and away from moisture. In solution, store at -80°C for 6 months or -20°C for 1 month.