Learn More
Cayman Chemical Tilmicosin (phosphate), Purity: ≥95%, CAS: 137330-13-3, 50 mg
SDP
Catalog No. 502965105
Encompass_Preferred
A macrolide antibiotic; active against macrolide-susceptible strains of M. haemolytica, P. multocida, and E. coli (MICs = 4, 4, and 128 µM, respectively), as well as a hyperpermeable strain of E. coli (MIC = 2 µM); inhibits bacterial protein synthesis (IC50 = 0.36 µM); increases serum CK, CK-MB, and MDA levels in mice, indicating cardiotoxicity
Catalog No. 50-296-5105 Supplier Cayman Chemical Supplier No. 1819050 MG
A macrolide antibiotic; active against macrolide-susceptible strains of M. haemolytica, P. multocida, and E. coli (MICs = 4, 4, and 128 µM, respectively), as well as a hyperpermeable strain of E. coli (MIC = 2 µM); inhibits bacterial protein synthesis (IC50 = 0.36 µM); increases serum CK, CK-MB, and MDA levels in mice, indicating cardiotoxicity