Learn More
TARGETMOL CHEMICALS INC TILOMISOLE 5MG
SDP
Catalog No. 502579550
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Tilomisole (Wy 18251) is an orally available benzimidazole thiazole with anti-inflammatory activity and immunomodulatory activity.Tilomisole is used in the study of cancer and inflammation. purity: 98%
Catalog No. 50-257-9550 Supplier TARGETMOL CHEMICALS INC Supplier No. T611075MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Tilomisole (Wy 18251) is an orally available benzimidazole thiazole with anti-inflammatory activity and immunomodulatory activity.Tilomisole is used in the study of cancer and inflammation. purity: 98%