Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tilorone Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. T2836100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2836100MG 100 mg
T28361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2836100MG Supplier TCI America Supplier No. T2836100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27591-69-1
Molecular Formula C25H36Cl2N2O3
Molecular Weight (g/mol) 483.47
MDL Number MFCD00134071
InChI Key BSVYJQAWONIOOU-UHFFFAOYSA-N
Synonym 2,7-Bis[2-(diethylamino)ethoxy]-9-fluorenone Dihydrochloride
PubChem CID 33958
IUPAC Name dihydrogen 2,7-bis[2-(diethylamino)ethoxy]-9H-fluoren-9-one dichloride
SMILES [H+].[H+].[Cl-].[Cl-].CCN(CC)CCOC1=CC=C2C3=CC=C(OCCN(CC)CC)C=C3C(=O)C2=C1

Specifications

Melting Point 234°C
Quantity 100 mg
Formula Weight 483.47
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tilorone Dihydrochloride
RTECSNumber LL9000000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.