Learn More
Cayman Chemical TIloronhydrochlorIde 100mg
SDP
Catalog No. 502965666
Encompass_Preferred
An orally active, antiviral compound that induces interferon production and is reported to have antitumor, anti-inflammatory, and neuroprotective actions
Catalog No. 50-296-5666 Supplier Cayman Chemical Supplier No. 17868100 MG
An orally active, antiviral compound that induces interferon production and is reported to have antitumor, anti-inflammatory, and neuroprotective actions