Learn More
MSC Tilsatec Cut Resistant Gloves, A4, 2X-Large, 12 Pairs
Catalog No. 5000675412
Encompass
These cut, puncture, and abrasion-resistant gloves are constructed with RhinoYarn and feature a polyurethane-coated palm and fingertips for a smooth grip. Designed for high protection, they are suitable for a range of industrial applications requiring superior hand safety.
  • Knit style
  • Polyurethane coating on palm and fingertips
  • ANSI/ISEA Cut Resistance Level A4
  • RhinoYarn primary material
  • ANSI/ISEA Abrasion Resistance Level 3
  • Lime color
  • Numeric size 11 (2X-Large)
  • ANSI/ISEA Puncture Resistance Level 3
  • Black palm coating color
  • EN 388 Rating: 3X42D
  • Seamless Rhino Yarn lining
  • Smooth grip surface
  • Knit cuff style
  • Unisex use
Catalog No. 50-006-75412 Supplier MSC Supplier No. 90691924
These cut, puncture, and abrasion-resistant gloves are constructed with RhinoYarn and feature a polyurethane-coated palm and fingertips for a smooth grip. Designed for high protection, they are suitable for a range of industrial applications requiring superior hand safety.
  • Knit style
  • Polyurethane coating on palm and fingertips
  • ANSI/ISEA Cut Resistance Level A4
  • RhinoYarn primary material
  • ANSI/ISEA Abrasion Resistance Level 3
  • Lime color
  • Numeric size 11 (2X-Large)
  • ANSI/ISEA Puncture Resistance Level 3
  • Black palm coating color
  • EN 388 Rating: 3X42D
  • Seamless Rhino Yarn lining
  • Smooth grip surface
  • Knit cuff style
  • Unisex use