Learn More
MSC Tilsatec Glove, ANSI Cut A6, Latex Palm, 1 Pair
Catalog No. 5000802890
Encompass
The TTP060L is an original latex palm-coated ANSI level A6 cut-resistant glove. Its crinkled latex coating on the palm and fingers provides excellent grip in wet applications and high levels of puncture protection, channeling surface liquid away for safe handling.
  • ANSI level A6 cut resistance
  • ANSI puncture level 4
  • Wrinkled latex coating on palm and fingers for excellent grip in wet applications and high puncture protection
Catalog No. 50-008-02890 Supplier MSC Supplier No. 65544488
The TTP060L is an original latex palm-coated ANSI level A6 cut-resistant glove. Its crinkled latex coating on the palm and fingers provides excellent grip in wet applications and high levels of puncture protection, channeling surface liquid away for safe handling.
  • ANSI level A6 cut resistance
  • ANSI puncture level 4
  • Wrinkled latex coating on palm and fingers for excellent grip in wet applications and high puncture protection