Learn More
MSC Tilsatec Gloves, Cut A4, Puncture 3, Size Medium, 12 Pair
Catalog No. 5000765611
Encompass
These Tilsatec Cut, Puncture & Abrasion-Resistant Gloves are engineered with RhinoYarn material and feature a polyurethane-coated palm and fingertips for enhanced grip and protection. Designed for industrial applications requiring high cut, puncture, and abrasion resistance, these medium-sized gloves offer a seamless knit style for comfortable wear.
  • Made with RhinoYarn primary material
  • Feature polyurethane coating on palm and fingertips for smooth grip
  • Rated ANSI/ISEA Cut Level A4, Puncture Resistance Level 3, and Abrasion Resistance Level 3
  • Seamless knit construction with knit cuff
  • Tear-away design for safety
  • Unisex design in size Medium (Numeric Size: 8)
  • Glove color is lime with black palm coating
Catalog No. 50-007-65611 Supplier MSC Supplier No. 90691767
These Tilsatec Cut, Puncture & Abrasion-Resistant Gloves are engineered with RhinoYarn material and feature a polyurethane-coated palm and fingertips for enhanced grip and protection. Designed for industrial applications requiring high cut, puncture, and abrasion resistance, these medium-sized gloves offer a seamless knit style for comfortable wear.
  • Made with RhinoYarn primary material
  • Feature polyurethane coating on palm and fingertips for smooth grip
  • Rated ANSI/ISEA Cut Level A4, Puncture Resistance Level 3, and Abrasion Resistance Level 3
  • Seamless knit construction with knit cuff
  • Tear-away design for safety
  • Unisex design in size Medium (Numeric Size: 8)
  • Glove color is lime with black palm coating