Learn More
MSC Tilsatec Safety Glove, A4 Cut/Puncture/Abrasion 3, X-Large, Tear-Away, 1 Pair
Catalog No. 5000688336
Encompass
The Tilsatec TTP333DT-PU CR+ DeTach™ safety glove provides extreme cut resistance and features built-in tear-away zones on the fingers. This innovative design helps prevent hands from being pulled into moving machinery by allowing the glove fingers to detach if trapped, minimizing injury risk while offering ANSI A4 cut protection.
  • ANSI A4 cut resistance (>1,500 grams)
  • CR+ DeTach™ tear-away technology with 5 built-in tear-away zones on the fingers
  • Fine gauge lightweight shell provides high tactility and dexterity
  • Polyurethane palm coating ensures good grip, durability, and protection
  • Open back design allows hands to breathe
  • Minimizes risk of traumatic finger injuries, broken bones, and amputations
Catalog No. 50-006-88336 Supplier MSC Supplier No. 90691486
The Tilsatec TTP333DT-PU CR+ DeTach™ safety glove provides extreme cut resistance and features built-in tear-away zones on the fingers. This innovative design helps prevent hands from being pulled into moving machinery by allowing the glove fingers to detach if trapped, minimizing injury risk while offering ANSI A4 cut protection.
  • ANSI A4 cut resistance (>1,500 grams)
  • CR+ DeTach™ tear-away technology with 5 built-in tear-away zones on the fingers
  • Fine gauge lightweight shell provides high tactility and dexterity
  • Polyurethane palm coating ensures good grip, durability, and protection
  • Open back design allows hands to breathe
  • Minimizes risk of traumatic finger injuries, broken bones, and amputations