Learn More
MSC Tilsatec Tear-Away Safety Glove, Cut A1, X-Small, 12 Pair
Catalog No. 5000792632
Encompass
Tilsatec's GP2000PU DeTach™ gloves feature an innovative hand protection concept with 14 built-in tear-away zones on the fingers, designed to minimize injury risk when working with moving machinery or rotational tools. These gloves also include a polyurethane palm coating for enhanced grip, durability, and protection, alongside an open back for breathability, making them a versatile choice for various applications.
  • A1 cut resistance per ANSI 105: 2016
  • De|Tach™ tear-away technology
  • Fine gauge lightweight shell for high tactility and dexterity
  • Features 14 built-in tear-away zones on fingers
  • Polyurethane palm coating provides good grip, durability, and protection
  • Open back allows hands to breathe
  • Seamless knit construction
  • Integrated knit wrist
Catalog No. 50-007-92632 Supplier MSC Supplier No. 90692153
Tilsatec's GP2000PU DeTach™ gloves feature an innovative hand protection concept with 14 built-in tear-away zones on the fingers, designed to minimize injury risk when working with moving machinery or rotational tools. These gloves also include a polyurethane palm coating for enhanced grip, durability, and protection, alongside an open back for breathability, making them a versatile choice for various applications.
  • A1 cut resistance per ANSI 105: 2016
  • De|Tach™ tear-away technology
  • Fine gauge lightweight shell for high tactility and dexterity
  • Features 14 built-in tear-away zones on fingers
  • Polyurethane palm coating provides good grip, durability, and protection
  • Open back allows hands to breathe
  • Seamless knit construction
  • Integrated knit wrist