Learn More
Grainger Ergonomic Tilt Back Cylinder
Catalog No. NC2881929
Encompass_Preferred
The Ergonomic Tilt Back Cylinder by Global Industrial is a pneumatic gas cylinder that provides tilt adjustment for chairs and furniture. It has a maximum weight capacity of 250 lbs and adjusts from 3.5 inches to 21 inches high. The cylinder is made from steel and aluminum construction with a black powder coated finish. It has a threaded female mount on bottom and male mount on top. The Ergonomic Tilt Back Cylinder requires 60-100 PSI to operate and allows the user to adjust the tilt tension. This product provides ergonomic support and custom comfort.
Catalog No. NC2881929 Supplier Grainger Supplier No. 19G945
The Ergonomic Tilt Back Cylinder by Global Industrial is a pneumatic gas cylinder that provides tilt adjustment for chairs and furniture. It has a maximum weight capacity of 250 lbs and adjusts from 3.5 inches to 21 inches high. The cylinder is made from steel and aluminum construction with a black powder coated finish. It has a threaded female mount on bottom and male mount on top. The Ergonomic Tilt Back Cylinder requires 60-100 PSI to operate and allows the user to adjust the tilt tension. This product provides ergonomic support and custom comfort.