Learn More
World Precision Instrument Tilting Base with Screw Adjustment
SDP
Catalog No. 50822977
Encompass
At last! A tilt base you can operate with one hand!
  • High precision and high stability
  • Tilt angle up to 35º
  • 3.5-lb steel base provides stability for your micromanipulator
  • Holes also allow permanent mount­ing to your bench top
  • Adjust tilt angle by turning knob
  • Manipulator mounting bracket included
Catalog No. 50-822-977 Supplier World Precision Instrument Supplier No. TBS
At last! A tilt base you can operate with one hand!
  • High precision and high stability
  • Tilt angle up to 35º
  • 3.5-lb steel base provides stability for your micromanipulator
  • Holes also allow permanent mount­ing to your bench top
  • Adjust tilt angle by turning knob
  • Manipulator mounting bracket included